missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-methyl-1,3-thiazole-4-carboxylate, 97%, Maybridge™

Product Code. 10263572
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10263572 1 g Amber Glass Bottle 1g
10682463 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10263572 Supplier Thermo Scientific Maybridge Supplier No. SPB02288DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6436-59-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00156148
InChI Key QWWPUBQHZFHZSF-UHFFFAOYSA-N
PubChem CID 293353
IUPAC Name ethyl 2-methyl-1,3-thiazole-4-carboxylate
SMILES CCOC(=O)C1=CSC(=N1)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym ethyl 2-methylthiazole-4-carboxylate, 2-methyl-thiazole-4-carboxylic acid ethyl ester, ethyl2-methylthiazole-4-carboxylate, 4-thiazolecarboxylic acid, 2-methyl-, ethyl ester, pubchem9870, maybridge1_008348, acmc-1b4zl, ksc495k3j, ethyl 2-methylthiazol-4-carboxylate, ethyl 2-methyl-4-thiazolecarboxylate
Formula Weight 171.22
Percent Purity 97%
Chemical Name or Material ethyle2-methyl-1, 3-thiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.