CAS RN 6436-59-5
IUPAC Name:
ethyl 2-methyl-1,3-thiazole-4-carboxylate
Synonyms:
ethyl 2-methyl-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol):
171.21
Molecular Formula:
C7H9NO2S
InChi Key:
QWWPUBQHZFHZSF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CSC(C)=N1
1
–
2
of
2
results
1
ethyle2-methyl-1,3-thiazole-4-carboxylate, 97%, Maybridge™
CAS: 6436-59-5 Molecular Formula: C7H9NO2S Molecular Weight (g/mol): 171.214 MDL Number: MFCD00156148 InChI Key: QWWPUBQHZFHZSF-UHFFFAOYSA-N PubChem CID: 293353 IUPAC Name: ethyl 2-methyl-1,3-thiazole-4-carboxylate SMILES: CCOC(=O)C1=CSC(=N1)C
2
Ethyl 2-methylthiazole-4-carboxylate, 98%, Thermo Scientific™
CAS: 6436-59-5 Molecular Formula: C7H9NO2S Molecular Weight (g/mol): 171.214 MDL Number: MFCD00156148 InChI Key: QWWPUBQHZFHZSF-UHFFFAOYSA-N PubChem CID: 293353 IUPAC Name: ethyl 2-methyl-1,3-thiazole-4-carboxylate SMILES: CCOC(=O)C1=CSC(=N1)C