missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylaluminum dichloride, 1M in hexanes, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mL
250 mL
Description
Ethylaluminum dichloride is used as a catalyst component in the polyolefin industry, for the synthesis of synthetic rubbers and for the dimerisation of olefins. It is also employed as a catalyst for Friedel-Crafts acylations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 0.729 g/mL |
Boiling Point | 68°C to 70°C |
Flash Point | -22°C (-9°F) |
Quantity | 10 mL |
UN Number | UN3394 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in acetone. Miscible with alcohol,chloroform,ether. Reacts with water. |
Formula Weight | 126.95 |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'