missing translation for 'onlineSavingsMsg'
Learn More

Ethylaluminum dichloride, 1M in hexanes

Product Code. 11326509
Change view
Click to view available options
Quantity:
10 mL
250 mL
Unit Size:
10mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326509 250 mL 250mL
11306509 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326509 Supplier Thermo Scientific Alfa Aesar Supplier No. 041712.AK

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol

Ethylaluminum dichloride is used as a catalyst component in the polyolefin industry, for the synthesis of synthetic rubbers and for the dimerisation of olefins. It is also employed as a catalyst for Friedel-Crafts acylations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-43-9
Molecular Formula C2H5AlCl2
Molecular Weight (g/mol) 126.94
MDL Number MFCD00000457
InChI Key UAIZDWNSWGTKFZ-UHFFFAOYSA-L
Synonym ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
IUPAC Name dichloro(ethyl)alumane
SMILES CC[Al](Cl)Cl

Specifications

Color Colorless to Yellow
Density 0.729 g/mL
Boiling Point 68°C to 70°C
Flash Point -22°C (-9°F)
Quantity 250 mL
UN Number UN3394
Sensitivity Air and moisture sensitive
Solubility Information Soluble in acetone. Miscible with alcohol,chloroform,ether. Reacts with water.
Formula Weight 126.95
Physical Form Liquid
Chemical Name or Material Ethylaluminum dichloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylaluminum dichloride, 1M in hexanes
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Self-heating substance Category 2
  • Substance which, in contact with water, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
  • H250-Catches fire spontaneously if exposed to air.
  • H252-Self-heating in large quantities; may catch fire.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P422-Store contents under .
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.