missing translation for 'onlineSavingsMsg'
Learn More

Ethyl xanthene-9-carboxylate, 98%, Thermo Scientific™

Product Code. 11498966
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498966 5 g 5g
11408976 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498966 Supplier Thermo Scientific Alfa Aesar Supplier No. A17095.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7401-03-8
ChEBI CHEBI:33344

Specifications

Melting Point 66°C to 68°C
Molecular Formula Nb
MDL Number MFCD00011126
Quantity 5 g
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES [Nb]
IUPAC Name niobium
Molecular Weight (g/mol) 92.91
PubChem CID 23936
Formula Weight 254.29
Percent Purity 98%
Chemical Name or Material Ethyl xanthene-9-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.