CAS RN 7401-03-8
IUPAC Name:
ethyl 9H-xanthene-9-carboxylate
Synonyms:
ethyl 9H-xanthene-9-carboxylate
Molecular Weight (g/mol):
254.29
Molecular Formula:
C16H14O3
InChi Key:
LFHPCJSHNRJCEY-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12
1
–
1
of
1
results
1
Ethyl xanthene-9-carboxylate, 98%, Thermo Scientific™
CAS: 7401-03-8 ChEBI: CHEBI:33344