missing translation for 'onlineSavingsMsg'
Learn More

Ethyl xanthene-9-carboxylate, 98%, Thermo Scientific™

Product Code. 11408976
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408976 25 g 25g
11498966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408976 Supplier Thermo Scientific Alfa Aesar Supplier No. A17095.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7401-03-8

Specifications

Melting Point 66°C to 68°C
Molecular Formula C16H14O3
MDL Number MFCD00100669
Quantity 25 g
Synonym ethyl xanthene-9-carboxylate, maybridge1_002093, 9h-xanthene-9-carboxylic acid ethyl ester
InChI Key LFHPCJSHNRJCEY-UHFFFAOYSA-N
SMILES CCOC(=O)C1C2=CC=CC=C2OC3=CC=CC=C13
IUPAC Name ethyl 9H-xanthene-9-carboxylate
Molecular Weight (g/mol) 254.29
PubChem CID 235688
Formula Weight 254.29
Percent Purity 98%
Chemical Name or Material Ethyl xanthene-9-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.