missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-3-acetate, 98%

Product Code. 11448587
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448587 100 g 100g
11428587 5 g 5g
11438587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448587 Supplier Thermo Scientific Alfa Aesar Supplier No. A19694.22
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 37784-63-7 | C8H10O2S | 170.23 g/mol

Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37784-63-7
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00005472
InChI Key FZBNQIWPYCUPAP-UHFFFAOYSA-N
Synonym ethyl thiophene-3-acetate, ethyl 3-thienylacetate, ethyl 3-thiopheneacetate, ethyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, ethyl ester, ethyl 3-thienylacetate #, acmc-1afw1, ethyl 2-3-thienyl acetate, bidd:gt0101, ethyl 2-thiophen-3-ylethanoate
PubChem CID 520865
SMILES CCOC(=O)CC1=CSC=C1

Specifications

Density 1.137
Boiling Point 110°C to 114°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.51
Quantity 100 g
Beilstein 122555
IUPAC Name ethyl 2-thiophen-3-ylacetate
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material Ethyl thiophene-3-acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.