accessibility menu, dialog, popup

UserName

H3COOS

CAS RN 37784-63-7

IUPAC Name: ethyl 2-(thiophen-3-yl)acetate
Synonyms: ethyl 2-(thiophen-3-yl)acetate
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: FZBNQIWPYCUPAP-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CSC=C1
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)