missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-S-(+)-Clopidogrel Sulfate, TRC

Product Code. 30380381
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380381 50 mg
30376330 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380381 Supplier Toronto Research Chemicals Supplier No. TRCE90410050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1357474-92-0
Molecular Formula C17H20ClNO6S2
Molecular Weight (g/mol) 433.93
Synonym (alphaS)-alpha-(2-Chlorophenyl)-6,7-dihydro-thieno[3,2-c]pyridine-5(4H)-acetic Acid Ethyl Ester Sulfate
SMILES CCOC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H18ClNO2S.H2O4S/c1-2-21-17(20)16(13-5-3-4-6-14(13)18)19-9-7-15-12(11-19)8-10-22-15;1-5(2,3)4/h3-6,8,10,16H,2,7,9,11H2,1H3;(H2,1,2,3,4)/t16-;/m0./s1
Chemical Name or Material Ethyl-S-(+)-Clopidogrel Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.