CAS RN 1357474-92-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl-S-(+)-Clopidogrel Sulfate, TRC
CAS: 1357474-92-0 Molecular Formula: C17H20ClNO6S2 Molecular Weight (g/mol): 433.93 Synonym: (alphaS)-alpha-(2-Chlorophenyl)-6,7-dihydro-thieno[3,2-c]pyridine-5(4H)-acetic Acid Ethyl Ester Sulfate SMILES: CCOC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O