missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenyl sulfone, 98+%, Thermo Scientific™

Product Code. p-7080948
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11413484

Brand: Thermo Scientific Alfa Aesar A10710.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-70-2
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.226
MDL Number MFCD00025081
InChI Key VBQUDDWATQWCPP-UHFFFAOYSA-N
Synonym ethyl phenyl sulfone, ethylsulfonyl benzene, ethylphenyl sulfone, ethyl phenyl sulphone, sulfone, ethyl phenyl, ethylsulphonyl benzene, ethylphenylsulfone, benzene, ethylsulfonyl, ethanesulfonyl benzene, 3blu
PubChem CID 69032
IUPAC Name ethylsulfonylbenzene
SMILES CCS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 40°C to 44°C
Density 1.14
Quantity 5 g
Beilstein 2045095
Formula Weight 170.23
Percent Purity ≥98%
Chemical Name or Material Ethyl phenyl sulfone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.