accessibility menu, dialog, popup

UserName

CAS RN 599-70-2

IUPAC Name: (ethanesulfonyl)benzene
Synonyms: ethyl phenyl sulfone
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: VBQUDDWATQWCPP-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results