missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenyl sulfone, 98+%, Thermo Scientific™

Product Code. 11433484
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433484 100 g 100g
11413484 5 g 5g
11423484 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433484 Supplier Thermo Scientific Alfa Aesar Supplier No. A10710.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-70-2
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.226
MDL Number MFCD00025081
InChI Key VBQUDDWATQWCPP-UHFFFAOYSA-N
Synonym ethyl phenyl sulfone, ethylsulfonyl benzene, ethylphenyl sulfone, ethyl phenyl sulphone, sulfone, ethyl phenyl, ethylsulphonyl benzene, ethylphenylsulfone, benzene, ethylsulfonyl, ethanesulfonyl benzene, 3blu
PubChem CID 69032
IUPAC Name ethylsulfonylbenzene
SMILES CCS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 40°C to 44°C
Density 1.14
Quantity 100 g
Beilstein 2045095
Formula Weight 170.23
Percent Purity ≥98%
Chemical Name or Material Ethyl phenyl sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.