missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N,N-dimethyloxamate, 98%

Product Code. 11382786
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382786 5 g 5g
11392786 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382786 Supplier Thermo Scientific Alfa Aesar Supplier No. L08923.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16703-52-9 | C6H11NO3 | 145.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16703-52-9
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00015153
InChI Key HMALWDVRMHVUAW-UHFFFAOYSA-N
Synonym ethyl n,n-dimethyloxamate, ethyl nn-dimethyloxamate, n,n-dimethyl ethyl oxamate, ethyl 2-dimethylamino-2-oxoacetate, acetic acid, dimethylamino oxo-, ethyl ester, ethyl dimethylcarbamoyl formate, oxamic acid, dimethyl-, ethyl ester, acetic acid, 2-dimethylamino-2-oxo-, ethyl ester, ethyl dimethylamino oxoacetate, aethyl-dimethyloxamat
PubChem CID 85553
IUPAC Name ethyl 2-(dimethylamino)-2-oxoacetate
SMILES CCOC(=O)C(=O)N(C)C

Specifications

Density 1.068
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.443
Quantity 5 g
Beilstein 1906209
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Ethyl N,N-dimethyloxamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.