CAS RN 16703-52-9
IUPAC Name:
ethyl (dimethylcarbamoyl)formate
Synonyms:
ethyl (dimethylcarbamoyl)formate
Molecular Weight (g/mol):
145.16
Molecular Formula:
C6H11NO3
InChi Key:
HMALWDVRMHVUAW-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)N(C)C
1
–
1
of
1
results
1
Ethyl N,N-dimethyloxamate, 98%
CAS: 16703-52-9 Molecular Formula: C6H11NO3 Molecular Weight (g/mol): 145.158 MDL Number: MFCD00015153 InChI Key: HMALWDVRMHVUAW-UHFFFAOYSA-N Synonym: ethyl n,n-dimethyloxamate,ethyl nn-dimethyloxamate,n,n-dimethyl ethyl oxamate,ethyl 2-dimethylamino-2-oxoacetate,acetic acid, dimethylamino oxo-, ethyl ester,ethyl dimethylcarbamoyl formate,oxamic acid, dimethyl-, ethyl ester,acetic acid, 2-dimethylamino-2-oxo-, ethyl ester,ethyl dimethylamino oxoacetate,aethyl-dimethyloxamat PubChem CID: 85553 IUPAC Name: ethyl 2-(dimethylamino)-2-oxoacetate SMILES: CCOC(=O)C(=O)N(C)C