missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-pyrimidinecarboxylate, 97%, Thermo Scientific™

Product Code. 10644185 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10644185 5 g Glass bottle 5g
10451674 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10644185 Supplier Thermo Scientific Acros Supplier No. 425940050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40929-50-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD03094043
InChI Key YCWDQAKDVQNVAR-UHFFFAOYSA-N
Synonym ethyl 5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, ethyl ester, pyrimidine-5-carboxylic acid ethyl ester, ethyl5-pyrimidinecarboxylate, 5-ethoxycarbonylpyrimidine, ethylpyrimidine-5-carboxylate, 2-ethylpyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid ethyl ester
PubChem CID 4223810
IUPAC Name ethyl pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=CN=C1

Specifications

Density 1.1600g/mL
Boiling Point 102°C (12.0 mmHg)
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.16
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Ethyl 5-pyrimidinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 5-pyrimidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.