accessibility menu, dialog, popup

UserName

H3COONN

CAS RN 40929-50-8

IUPAC Name: ethyl pyrimidine-5-carboxylate
Synonyms: ethyl pyrimidine-5-carboxylate
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: YCWDQAKDVQNVAR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results