missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-phenyloxazole-2-carboxylate, TRC

Product Code. 30381045
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381045 25 mg
30381231 250 mg
30376278 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381045 Supplier Toronto Research Chemicals Supplier No. TRCE93879525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13575-16-1
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.22
SMILES CCOC(=O)c1oc(cn1)c2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H11NO3/c1-2-15-12(14)11-13-8-10(16-11)9-6-4-3-5-7-9/h3-8H,2H2,1H3
Formula Weight 217.0739
Chemical Name or Material Ethyl 5-phenyloxazole-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.