CAS RN 13575-16-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 5-phenyloxazole-2-carboxylate, TRC
CAS: 13575-16-1 Molecular Formula: C12H11NO3 Molecular Weight (g/mol): 217.22 SMILES: CCOC(=O)c1oc(cn1)c2ccccc2