missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Biphenylacetate, TRC

Product Code. 30387801
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387801 500 mg -
30375494 50 mg 50mg
30387280 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387801 Supplier Toronto Research Chemicals Supplier No. TRCE900208500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14062-23-8
Molecular Formula C16 H16 O2
Molecular Weight (g/mol) 240.3
Synonym [1,1'-Biphenyl]-4-acetic acid ethyl ester, 4-Biphenylacetic acid, ethyl ester (8CI), Acetic acid, (4-biphenylyl)-, ethyl ester (7CI), Diatec, Ethyl 4-biphenylylacetate, Ethyl p-biphenylylacetate, Felbinac ethyl ester, LM 001
IUPAC Name ethyl 2-(4-phenylphenyl)acetate
SMILES CCOC(=O)Cc1ccc(cc1)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H16O2/c1-2-18-16(17)12-13-8-10-15(11-9-13)14-6-4-3-5-7-14/h3-11H,2,12H2,1H3
Formula Weight 240.115
Chemical Name or Material Felbinac Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.