CAS RN 14062-23-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ethyl 4-Biphenylacetate, TRC
CAS: 14062-23-8 Molecular Formula: C16 H16 O2 Molecular Weight (g/mol): 240.3 Synonym: [1,1'-Biphenyl]-4-acetic acid ethyl ester,4-Biphenylacetic acid, ethyl ester (8CI),Acetic acid, (4-biphenylyl)-, ethyl ester (7CI),Diatec,Ethyl 4-biphenylylacetate,Ethyl p-biphenylylacetate,Felbinac ethyl ester,LM 001 IUPAC Name: ethyl 2-(4-phenylphenyl)acetate SMILES: CCOC(=O)Cc1ccc(cc1)c2ccccc2
2
Felbinac Ethyl Ester, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.