missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxo-4-(2,4,5-trifluorophenyl)butanoate, TRC

Product Code. 30387620
Change view
Click to view available options
Quantity:
100 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387620 100 mg
30386054 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387620 Supplier Toronto Research Chemicals Supplier No. TRCE925830100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1151240-88-8
Molecular Formula C12H11F3O3
Molecular Weight (g/mol) 260.21
Synonym Ethyl Ester 2,4,5-Trifluoro-beta-oxo-benzenebutanoic Acid
SMILES CCOC(=O)CC(=O)Cc1cc(F)c(F)cc1F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H11F3O3/c1-2-18-12(17)5-8(16)3-7-4-10(14)11(15)6-9(7)13/h4,6H,2-3,5H2,1H3
Formula Weight 260.066
Chemical Name or Material Ethyl 3-Oxo-4-(2,4,5-trifluorophenyl)butanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.