CAS RN 1151240-88-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 3-Oxo-4-(2,4,5-trifluorophenyl)butanoate, TRC
CAS: 1151240-88-8 Molecular Formula: C12H11F3O3 Molecular Weight (g/mol): 260.21 Synonym: Ethyl Ester 2,4,5-Trifluoro-beta-oxo-benzenebutanoic Acid SMILES: CCOC(=O)CC(=O)Cc1cc(F)c(F)cc1F