missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (3-bromo-1H-pyrazol-1-yl)acetate, TRC

Product Code. 30388136
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388136 250 mg
30377380 25 mg
30383520 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388136 Supplier Toronto Research Chemicals Supplier No. TRCE901650250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1427022-51-2
Molecular Formula C7H9N2O2Br
Molecular Weight (g/mol) 233.06
Synonym 3-bromo-1H-Pyrazole-1-acetic Acid Ethyl Ester
SMILES CCOC(=O)Cn1ccc(Br)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H9BrN2O2/c1-2-12-7(11)5-10-4-3-6(8)9-10/h3-4H,2,5H2,1H3
Formula Weight 231.9847
Chemical Name or Material Ethyl (3-bromo-1H-pyrazol-1-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.