CAS RN 1427022-51-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl (3-bromo-1H-pyrazol-1-yl)acetate, TRC
CAS: 1427022-51-2 Molecular Formula: C7H9N2O2Br Molecular Weight (g/mol): 233.06 Synonym: 3-bromo-1H-Pyrazole-1-acetic Acid Ethyl Ester SMILES: CCOC(=O)Cn1ccc(Br)n1