missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(4-pyridyl)thiazole-4-acetate, 97%, Thermo Scientific™

Product Code. 15440548
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440548 250 mg 250mg
15420548 1 g 1g
15430548 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440548 Supplier Thermo Scientific Alfa Aesar Supplier No. H54192.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80653-68-5
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD06739821
InChI Key JZBJUBBYJUUKQK-UHFFFAOYSA-N
Synonym ethyl 2-2-pyridin-4-yl thiazol-4-yl acetate, ethyl 2-4-pyridyl thiazole-4-acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethyl ester, 2-4-pyridyl thiazole-4-acetic acid ethyl ester, ethyl 2-2-pyridin-4-yl-1,3-thiazol-4-yl acetate, 2-pyridin-4-yl-thiazol-4-yl-acetic acid ethylester
PubChem CID 7176350
IUPAC Name ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2

Specifications

Quantity 250 mg
Formula Weight 248.3
Percent Purity 97%
Chemical Name or Material Ethyl 2-(4-pyridyl)thiazole-4-acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.