CAS RN 80653-68-5
IUPAC Name:
ethyl 2-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]acetate
Synonyms:
ethyl 2-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]acetate
Molecular Weight (g/mol):
248.3
Molecular Formula:
C12H12N2O2S
InChi Key:
JZBJUBBYJUUKQK-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=CSC(=N1)C1=CC=NC=C1
1
–
1
of
1
results
1
Ethyl 2-(4-pyridyl)thiazole-4-acetate, 97%, Thermo Scientific™
CAS: 80653-68-5 Molecular Formula: C12H12N2O2S Molecular Weight (g/mol): 248.3 MDL Number: MFCD06739821 InChI Key: JZBJUBBYJUUKQK-UHFFFAOYSA-N Synonym: ethyl 2-2-pyridin-4-yl thiazol-4-yl acetate,ethyl 2-4-pyridyl thiazole-4-acetate,2-pyridin-4-yl-thiazol-4-yl-acetic acid ethyl ester,2-4-pyridyl thiazole-4-acetic acid ethyl ester,ethyl 2-2-pyridin-4-yl-1,3-thiazol-4-yl acetate,2-pyridin-4-yl-thiazol-4-yl-acetic acid ethylester PubChem CID: 7176350 IUPAC Name: ethyl 2-(2-pyridin-4-yl-1,3-thiazol-4-yl)acetate SMILES: CCOC(=O)CC1=CSC(=N1)C2=CC=NC=C2