missing translation for 'onlineSavingsMsg'
Learn More

Ethanol-d, for NMR, 99.5+ atom % D

Product Code. 10387770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387770 100 g Glass Bottle 100g
10011940 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10387770 Supplier Thermo Scientific Acros Supplier No. 166321000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 925-93-9 | C2H6O | 47.075 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-93-9
Molecular Formula C2H6O
Molecular Weight (g/mol) 47.075
MDL Number MFCD00044669
InChI Key LFQSCWFLJHTTHZ-WFVSFCRTSA-N
Synonym ethanol-d, ethanol-d1, deuteroethanol, ethyl alcohol-d, ethyl 2 alcohol, ethanol-od, etod, ethan ol-d, ethanol-d 9ci, o-2h ethanol
PubChem CID 123093
IUPAC Name deuteriooxyethane
SMILES CCO

Specifications

Physical Form Liquid
Color Colorless
Density 0.8000g/mL
Deuterium (D) 99.50% min.
Packaging Glass Bottle
Water 0.5% max. (Coulometric)
Quantity 100 g
Linear Formula C2H5OD
Formula Weight 47.07
Grade NMR
Chemical Name or Material Ethanol-d, 99.5+ atom % D
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol-d
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.