accessibility menu, dialog, popup

UserName

CAS RN 925-93-9

CH3O2H

CAS RN 925-93-9

IUPAC Name: ethan(²H)ol
Synonyms: ethan(²H)ol
Molecular Weight (g/mol): 47.08
Molecular Formula: C2H6O
InChi Key: LFQSCWFLJHTTHZ-WFVSFCRTSA-N
SMILES: [2H]OCC
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results