Learn More
Estrogen Receptor Ab-17 Rabbit Polyclonal Antibody, Epredia™
Epredia™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody provides accurate, reproducible results.
£151.00 - £702.00
Specifications
| Antigen | Estrogen Receptor Ab-17 |
|---|---|
| Concentration | 1mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Purification Method | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Purification Method | Price | Quantity & Availability | ||||
|
12603017
|
Epredia
RB-1521-P0 |
100 μL | Purified with BSA and azide | ||||||
|
12613017
|
Epredia
RB-1521-P1 |
500 μL | Purified with BSA and azide | ||||||
|
12693007
|
Epredia
RB-1521-P |
1 mL | Purified with BSA and azide | ||||||
|
12623017
|
Epredia
RB-1521-P1ABX |
500 μL | Purified w/o BSA and azide | ||||||
|
12633017
|
Epredia
RB-1521-PABX |
1 mL | Purified w/o BSA and azide | ||||||
Beschreibung
The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein
Molecular Weight: 67 kDa
Positive Control: MCF-7 cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Spezifikation
| Estrogen Receptor Ab-17 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Steroids Hormones and Growth Factors | |
| A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein | |
| Primary |
| 1mg/mL | |
| Polyclonal | |
| Concentrated | |
| RUO | |
| Bovine, Human, Mouse, Pig, Rabbit, Rat | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.