missing translation for 'onlineSavingsMsg'
Learn More

Estrogen Receptor Ab-17 Rabbit Polyclonal Antibody, Epredia™

Epredia™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia RB-1521-P1

 View more versions of this product

Product Code. 12613017

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

ER gene consists of more than 140kb of genomic DNA divided into 8 exons, being translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Estrogen Receptor Ab-17
Polyclonal
1mg/mL
Rabbit
Purified with BSA and azide
RUO
Primary
Concentrated
Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein
500 μL
Steroids Hormones and Growth Factors
Bovine, Human, Mouse, Pig, Rabbit, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.