missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol Antibody (AB5.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£159.00
Specifications
| Antigen | Estradiol |
|---|---|
| Clone | AB5.1 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
Description
Estradiol Monoclonal specifically detects Estradiol in Mouse, Non-species specific samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.Specifications
| Estradiol | |
| 1 mg/mL | |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| RUO | |
| Estradiol-6-CMO-KLH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| AB5.1 | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Monoclonal | |
| Mouse | |
| Other | |
| IgG1 | |
| Affinity Purified | |
| Chemical. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title