missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol Antibody (AB5.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-78621-0.025mg
This item is not returnable.
View return policy
Description
Estradiol Monoclonal specifically detects Estradiol in Mouse, Non-species specific samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.
Specifications
| Estradiol | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Estradiol-6-CMO-KLH | |
| 0.025 mg | |
| Primary | |
| Other | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| AB5.1 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Chemical. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction