missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESAM Rabbit anti-Human, Clone: 113, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89321-100UL
This item is not returnable.
View return policy
Description
ESAM Monoclonal specifically detects ESAM in Human samples. It is validated for Flow Cytometry.
Specifications
| ESAM | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 90952 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry | |
| 113 | |
| Flow Cytometry 1:25-1:100 | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| Recombinant Human ESAM protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction