missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESAM Rabbit anti-Human, Clone: 113, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | ESAM |
|---|---|
| Clone | 113 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261176
|
Novus Biologicals
NBP2-89321-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18231916
|
Novus Biologicals
NBP2-89321-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ESAM Monoclonal specifically detects ESAM in Human samples. It is validated for Flow Cytometry.Specifications
| ESAM | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 90952 | |
| IgG | |
| Protein A purified |
| 113 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| Recombinant Human ESAM protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title