missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERVFRD-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | ERVFRD-1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18323047
|
Novus Biologicals
NBP3-04761-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18317874
|
Novus Biologicals
NBP3-04761-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERVFRD-1 Polyclonal specifically detects ERVFRD-1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| ERVFRD-1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 405754 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| endogenous retrovirus group FRD member 1, endogenous retrovirus group FRD, member 1, envelope polyprotein, envFRD, ERVFRDE1, FLJ41944, GLLL6191, HERV-FRD, HERV-FRD provirus ancestral Env polyprotein, HERV-FRD_6p24.1 provirus ancestral Env polyprotein, HERV-W/FRD, MGC87585, syncytin 2, UNQ6191, UNQ6191/PRO20218 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 16-250 of human ERVFRD-1 (NP_997465.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title