missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERVFRD-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04761-100UL
This item is not returnable.
View return policy
Description
ERVFRD-1 Polyclonal specifically detects ERVFRD-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ERVFRD-1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 405754 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| endogenous retrovirus group FRD member 1, endogenous retrovirus group FRD, member 1, envelope polyprotein, envFRD, ERVFRDE1, FLJ41944, GLLL6191, HERV-FRD, HERV-FRD provirus ancestral Env polyprotein, HERV-FRD_6p24.1 provirus ancestral Env polyprotein, HERV-W/FRD, MGC87585, syncytin 2, UNQ6191, UNQ6191/PRO20218 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 16-250 of human ERVFRD-1 (NP_997465.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction