missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERMAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ERMAP |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ERMAP Polyclonal specifically detects ERMAP in Human samples. It is validated for Western Blot.Specifications
| ERMAP | |
| Polyclonal | |
| Rabbit | |
| Q96PL5 | |
| 114625 | |
| IgG | |
| 52 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RDMGC118812, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813 | |
| ERMAP | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title