missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERMAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62513
This item is not returnable.
View return policy
Description
ERMAP Polyclonal specifically detects ERMAP in Human samples. It is validated for Western Blot.
Specifications
| ERMAP | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RDMGC118812, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813 | |
| Rabbit | |
| 52 kDa | |
| 100 μL | |
| Primary | |
| Dog: 79%. | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96PL5 | |
| ERMAP | |
| Synthetic peptides corresponding to ERMAP(erythroblast membrane-associated protein (Scianna blood group)) The peptide sequence was selected from the middle region of ERMAP. Peptide sequence PANGHWLLRQSRGNEYEALTSPQTSFRLKEPPRCVGIFLDYEAGVISFYN The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 114625 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction