missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Erlenmeyer Shaker Flasks with Flat Bottom

Ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. Each flask is manufactured utilizing an optically clear, inert polycarbonate resin which can be disposed of after single use or be made reusable when autoclaved.

Brand:  Greiner Bio-One 679502

Additional Details : Weight : 1.10000kg

 View more versions of this product

Product Code. 15944462

  • £118.08 / Pack of 12

Estimated Shipment: 03-06-2022
Add to basket

Description

Description

  • 2-in-1 DuoCAP™ for flexibility of sterile air exchange
    or tight seal
  • Made of optically clear polycarbonate which is
    lighter and safer than glass
  • Individually packaged and sterile to SAL 10-6 for
    long term culture applications
  • Molded-in graduations for easy volumetric reference
  • Non-pyrogenic, non-cytotoxic, and nuclease-free
  • Available with flat or baffled base for increased
    agitation and aeration
Specifications

Specifications

Clear
Polycarbonate
142.3 mm (w/cap)
Sterile
250 mL
Suspension culture
83 mm base
Flat
Videos
SDS
Documents
Certificates
View accessory products
Special Offers

Special Offers