missing translation for 'onlineSavingsMsg'
Learn More

Eritadenine, TRC

Product Code. 30378605
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378605 25 mg
30371059 10 mg
30381904 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378605 Supplier Toronto Research Chemicals Supplier No. TRCE60010025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23918-98-1
Molecular Formula C9H11N5O4
Molecular Weight (g/mol) 253.21
Synonym (αR,βR)-6-Amino-α,β-dihydroxy-9H-purine-9-butanoic Acid, 4-(6-Amino-9H-purin-9-yl)-4-deoxy-D-erythronic Acid, 4-(6-Aminopurin-9-yl)-4-deoxy-D-erythronic Acid, Lentinacin, Lentysine, 4-(9-Adenyl)-D-erythro-2,3-dihydroxybutyric Acid, D-Eritadenine
SMILES C1=NC2=C(C(=N1)N)N=CN2C[C@H]([C@H](C(=O)O)O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H11N5O4/c10-7-5-8(12-2-11-7)14(3-13-5)1-4(15)6(16)9(17)18/h2-4,6,15-16H,1H2,(H,17,18)(H2,10,11,12)/t4-,6-/m1/s1
Percent Purity >95%
Chemical Name or Material Eritadenine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.