CAS RN 23918-98-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Eritadenine, TRC
CAS: 23918-98-1 Molecular Formula: C9H11N5O4 Molecular Weight (g/mol): 253.21 Synonym: (αR,βR)-6-Amino-α,β-dihydroxy-9H-purine-9-butanoic Acid,4-(6-Amino-9H-purin-9-yl)-4-deoxy-D-erythronic Acid,4-(6-Aminopurin-9-yl)-4-deoxy-D-erythronic Acid,Lentinacin,Lentysine,4-(9-Adenyl)-D-erythro-2,3-dihydroxybutyric Acid,D-Eritadenine SMILES: C1=NC2=C(C(=N1)N)N=CN2C[C@H]([C@H](C(=O)O)O)O