Learn More
Macherey-Nagel™ Ergonomic Crimper
Product Code. 12614577 Shop All Macherey-Nagel Products
Click to view available options
Material:
Aluminum
Unit Size:
Each
Product Code. 12614577
Brand: Macherey-Nagel™ 735220
Only null left
Add to basket
Add to basket
Specifications
Product Type Ergonomic Crimper
For Use With (Equipment) 20 mm Aluminum Crimp Cap
Material Aluminum
No. per Pack 1 Ea.