missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ERCC8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ERCC8 Polyclonal specifically detects ERCC8 in Human samples. It is validated for Western Blot.Specifications
| ERCC8 | |
| Polyclonal | |
| Rabbit | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| Q13216 | |
| 1161 | |
| IgG | |
| 44 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8 | |
| ERCC8 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title