missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54898
This item is not returnable.
View return policy
Description
ERCC8 Polyclonal specifically detects ERCC8 in Human samples. It is validated for Western Blot.
Specifications
| ERCC8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8 | |
| Rabbit | |
| 44 kDa | |
| 100 μL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 1161 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q13216 | |
| ERCC8 | |
| Synthetic peptides corresponding to ERCC8(excision repair cross-complementing rodent repair deficiency, complementation group 8) The peptide sequence was selected from the middle region of ERCC8. Peptide sequence FQELYSGSRDCNILAWVPSLYEPVPDDDETTTKSQLNPAFEDAWSSSDEE The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction