missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ERC1 |
|---|---|
| Concentration | 0.47 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16831745
|
Proteintech
22211-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16821745
|
Proteintech
22211-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF-4 is a signal transducer associated with the cytoplasmic domain of the 75 kDa tumor necrosis factor receptor (TNF-R2).Specifications
| ERC1 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8IUD2 | |
| 23085 | |
| ERC1 Fusion Protein Ag17536 | |
| Primary | |
| -20°C | |
| ERC1 |
| 0.47 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cast2, ELKS, ERC 1, ERC1, KIAA1081, Rab6 interacting protein 2, RAB6IP2 | |
| ERC1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title