missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22211-1-AP-20UL
This item is not returnable.
View return policy
Description
TRAF-4 is a signal transducer associated with the cytoplasmic domain of the 75 kDa tumor necrosis factor receptor (TNF-R2).Specifications
| ERC1 | |
| Polyclonal | |
| Unconjugated | |
| ERC1 | |
| Cast2, ELKS, ERC 1, ERC1, KIAA1081, Rab6 interacting protein 2, RAB6IP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23085 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IUD2 | |
| ERC1 | |
| ERC1 Fusion Protein Ag17536 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction