missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Erbstatin Analog

Product Code. 15404189
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 15404189

Brand: Thermo Scientific Chemicals J61612.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An inhibitor of the EGF receptor associated tyrosine kinase

An inhibitor of the EGF receptor associated tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63177-57-1
Molecular Formula C10H10O4, C10H10O4
Synonym Methyl 2,5-dihydroxycinnamate

Specifications

Quantity 10 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) An inhibitor of the EGF receptor associated tyrosine kinase
Formula Weight 194.19g/mol
Solubility Soluble in methanol or 0.1M NaOH; Insoluble in water
Product Type Signal Transduction Reagent
Form Powder
InChI Key BQCNSTFWSKOWMA-GORDUTHDSA-N
SMILES COC(=O)\C=C\C1=CC(O)=CC=C1O
IUPAC Name methyl (2E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol) 194.19
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.