missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ErbB3/Her3 [p Tyr1289] Antibody (0004), Novus Biologicals™
Rabbit Monoclonal Antibody
£148.00 - £285.00
Specifications
| Antigen | ErbB3/Her3 |
|---|---|
| Clone | 0004 |
| Dilution | Western Blot 1:2000-1:20000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30042573
|
Novus Biologicals™
NBP3-30737-100ul |
100 μL |
£285.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30042700
|
Novus Biologicals™
NBP3-30737-20ul |
20 μL |
£148.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Western BlotSpecifications
| ErbB3/Her3 | |
| Western Blot 1:2000-1:20000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Tyr1289 of the Human ErbB3/Her3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 0004 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 0.2 μm filtered solution in PBS | |
| 2065 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title